VION Water, ACS Reagent Grade

Supplier: ALDON CORP SE

VN-WW7000-125ML VN-WW7000-1L VN-WW7000-250ML VN-WW7000-500ML VN-WW7000-60ML
470358-616EA 14.77 CAD
470358-616 470358-618 470358-612 470358-602 470358-606
VION Water, ACS Reagent Grade
Chemicals for Science Education

CAS Number: 7732-18-5
Formula Weight: 18.02
Formula: H2O

Guaranteed to meet the purity standards set by the American Chemical Society.



  • Reverse osmosis treated and deionized (RODI) water

  • Ideal for sensitive analytical analyses like HPLC.


Vion ACS Reagent grade water is guaranteed to meet the purity standards set by the American Chemical Society (ACS). It is a high-quality, highly purified water with minimal contaminants, and is ideal for use in laboratory settings for sensitive chemical analyses, like liquid chromatography and UV-Visible spectrophotometry, and reactions where extreme purity is required.



Specifications
Conductivity ≤ 2.0 μS/cm
Silicate (as SiO2) ≤ 0.01 ppm
Heavy Metals (as Pb) ≤ 0.01 ppm
Substances Reducing Permanganate - Passes Test
Chloride (Cl) ≤ 0.4 ppm
Nitrate (NO3) ≤ 0.4 ppm
Phosphate (PO4) ≤ 1 ppm
Sulfate (SO4) ≤ 1 ppm
UV A300 nm ≤ 0.005
UV A254 nm ≤ 0.005
UV A210 nm ≤ 0.010
UV A200 nm ≤ 0.010
LC Absorbance - Passes Test
LC Gradient Elution - Passes Test


Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR