Epic High Performance Cleanroom Coveralls
Supplier: TIANS INTERNATIONAL INC. TE
226853-2XL 226853-3XL 226853-4XL 226853-5XL 226853-L 226853-M 226853-XL
77938-772CS 161.16 CAD
77938-772 77938-766 77938-762 77938-768 77938-764 77938-770 77938-774
Epic High Performance Cleanroom Coveralls
Disposable Garments
Latex-free/lint-free, film-coated coverall designed for use in critical and low particulate clean environments. Fabric is suitable for exposure to bloodborne pathogens, some hazardous liquids, and where liquid splash is a concern.
  • Attached hood
  • Elastic wrist and ankles
  • Serged seams
  • Helmk drum tested
  • ISO Class 6
  • Latex-free
Fabric is constructed from a blend of polyethylene and polypropylene into a unique breathable membrane. The finished fabric also contains anti-static properties from a proprietary additive. Garments are individually inspected for sizing, stitching, and workmanship.
Features include an attached hood, zipper front entry, elastic wrists, elastic ankles, elastic lower back, and a storm flap zipper cover. Serged seam construction.
Packaging:
Coveralls are packaged 25 per case.