Cooled Incubators, KB Pro
Supplier: VWR International
77938-516 77938-518 77938-520 77938-522 77938-524 77938-526
77938-516EA 22840.46 CAD
77938-516 77938-518 77938-520 77938-522 77938-524 77938-526
Cooled Incubators, KB Pro
Incubators Cooled Incubators
A temperature control cabinet that can do it all, the KB PRO is our most powerful cooling incubator. With a temperature range of –20 to +100 °C and comprehensive programming options, it covers a wide range of applications that extend well beyond incubation.
  • Reliable and powerful thanks to innovative temperature control technology
  • Failsafe operation over the long term thanks to proven compressor technology and inner chamber made of stainless steel
  • Signal triangle with light-up status display, ergonomic control terminal with touchscreen program controller, rack with telescopic rails
  • Low energy consumption thanks to modern inverter cooling technology
  • Door heating to prevent condensation
  • Adjustable fan speed
  • Stainless-steel inner chamber and large access port with silicone plug
  • Inner doors made of metal oxide-coated safety glass
Two racks made of stainless steel (for models from 260 with telescopic rails). Tabletop units are stackable with stacking adapter. Intuitive touchscreen controller with time-segment and real-time programming.
Ergonomically adjustable control terminal. Floor-standing units featuring robust twin casters, with brakes at the front. Signal triangle with light-up status display. Class 3.3 independent temperature safety device (DIN 12880).