Cryo Hold Labels
Supplier: Computype
18904201 18904202 18904203 18904204
77591-848KT 802.69 CAD
77591-848 77591-850 77591-852 77591-854
Cryo Hold Labels
Labels
Eco-conscious label materials for sustainable practices.
  • 25% post consumer waste label material
  • Recyclable liners help to minimize harmful waste
  • Eco-friendly emulsion adhesives reduce environmental impact
Engineered to withstand temperatures down to –80 °C, these polyester labels feature a strong permanent adhesive with a high initial tack and a matte finish. Developed for thermal transfer printing, they include a compatible thermal transfer ribbon.
Certifications:
Computype collaborates with EcoVadis to manage and achieve sustainability goals.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable