VWR® Basic Industrial Hoods with Integrated Face Mask
Supplier: VWR International
77870-954 77870-956
77870-954CS 205.08 CAD
77870-954 77870-956
VWR® Basic Industrial Hoods with Integrated Face Mask
Disposable Garments
The VWR® Basic Industrial hood with integrated face mask is a disposable hood made from non-woven, spunbonded polypropylene (SPP) fabric. Featuring a two-ply face mask, it provides protection against dirt, grime, and certain dry particulates in non-hazardous environments, making it ideal for general industrial applications.
  • Free from natural rubber latex
  • Available in white or blue
  • Two-ply face mask is integrated into the hood
The VWR® Basic Industrial hood with integrated face mask is a lightweight, disposable solution designed for use in non-hazardous environments. Constructed from breathable, non-woven spunbonded polypropylene (SPP), it offers reliable protection against dust, dirt, and dry particulates. The integrated two-ply face mask enhances coverage, ensuring an added barrier against contaminants while maintaining comfort for extended wear. With an elasticized face opening for a secure fit and a durable design, this hood is ideal for industrial settings, clean manufacturing, food processing, and other applications where particulate control is essential.
Packaging:
Hoods are packaged 50 pieces per bag, 10 bags per case.