VWR® Contour™ Mobile Cabinets, Avantor
Supplier: VWR International
CFD-1812-B41 CFD-1814-B41 CFD-1842-B41 CFD-1844-B41 CFE-1402-B41 CFE-1402L-B41 CFE-1404-B41 CFE-1404L-B41 CFE-1405-B41 CFE-1416-B41 CWD-1812-B41 CWD-1814-B41 CWD-1842-B41 CWD-1844-B41 CWE-1402-B41 CWE-1402L-B41 CWE-1404-B41 CWE-1404L-B41 CWE-1405-B41 CWE-1416-B41 CWC-1032-B41 CWC-1032L-B41 CWC-1034-B41 CWC-1034L-B41 CWC-1035-B41 CWC-1036-B41 CWD-1002-B41 CWD-1004-B41 CWD-1005-B41 CWD-1006-B41 CFC-1032-B41 CFC-1032L-B41 CFC-1034-B41 CFC-1034L-B41 CFC-1035-B41 CFC-1036-B41 CFD-1002-B41 CFD-1004-B41 CFD-1005-B41 CFD-1006-B41
CFD-1812-B41EA 2531.08 CAD
CFD-1812-B41 CFD-1814-B41 CFD-1842-B41 CFD-1844-B41 CFE-1402-B41 CFE-1402L-B41 CFE-1404-B41 CFE-1404L-B41 CFE-1405-B41 CFE-1416-B41 CWD-1812-B41 CWD-1814-B41 CWD-1842-B41 CWD-1844-B41 CWE-1402-B41 CWE-1402L-B41 CWE-1404-B41 CWE-1404L-B41 CWE-1405-B41 CWE-1416-B41 CWC-1032-B41 CWC-1032L-B41 CWC-1034-B41 CWC-1034L-B41 CWC-1035-B41 CWC-1036-B41 CWD-1002-B41 CWD-1004-B41 CWD-1005-B41 CWD-1006-B41 CFC-1032-B41 CFC-1032L-B41 CFC-1034-B41 CFC-1034L-B41 CFC-1035-B41 CFC-1036-B41 CFD-1002-B41 CFD-1004-B41 CFD-1005-B41 CFD-1006-B41
VWR® Contour™ Mobile Cabinets, Avantor
Furniture Storage Cabinets
American-made, steel mobile cabinets, shipped fully assembled.
  • Numerous sizes and configurations
  • Two color options
  • Ships fully assembled
  • 3" Swivel locking caster
  • Powder-coated
The VWR® Contour™ mobile cabinet line is designed to integrate seamlessly into your existing Contour™ lab design, with the integrated flush pull design. Optioned in both light neutral and snow white powder coat, this cold rolled steel units come fully assembled and ready for use right out of the box.
Certifications:
Complies with SEFA 10 requirements.
Delivery information:
Fully assembled.
Packaging:
Boxed and palletized.