Cleanroom Folded Gown Dispensers
Supplier: PALBAM 2008 AC LTD TE
GD-0301 GD-0404
77898-362EA 1647.18 CAD
77898-362 77898-352
Cleanroom Folded Gown Dispensers
PPE Dispensers
Palbam’s folded gown dispenser is a wall mountable, stainless steel unit designed for cleanrooms. It ensures easy, contamination-free access to folded gowns, promoting organisation and compliance.
  • Stainless steel 304 with electropolished finish
  • Wall mountable
The gownroom is often the first impression for employees and visitors alike. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all ergonomic, cleanliness, and functional requirements.