SuperSlik® Low Retention Pipette Tips
Supplier: Labcon
1065-260-000-9 1065-960-008-9 1161-260-000-9 1161-960-008-9 1161-965-008-9 1165-960-008-9 1065-965-008-9 1066-260-000-9 1066-960-008-9 1066-965-008-9 1067-260-000-9 1067-960-008-9 1067-965-008-9 1068-260-000-9 1068-960-008-9 1068-965-008-9 1165-965-008-9 1168-260-000-9 1168-960-008-9 1168-965-008-9
77871-190CS 485.61 CAD
77871-190 77871-192 77871-246 77871-248 77871-250 77871-252 77871-194 77871-196 77871-198 77871-200 77871-202 77871-204 77871-206 77871-208 77871-210 77871-212 77871-254 77871-256 77871-258 77871-260
SuperSlik® Low Retention Pipette Tips
Pipette Tips
Labcon's ZAP™ aerosol filter pipette tips have been proven with case studies to block aerosol contamination. Unlike some leading brands of self-sealing aerosol-resistant pipette tips, ZAP™ tips have no cellulose gum, toluene solvent in the filter to mix with and contaminate the sample. ZAP™ tips will not wick fluids like cellulose-filled aerosol-resistant tips.
  • Patented 10 µm, hydrophobic filter blocks aerosols, preventing cross-contamination
  • Uniquely engineered to fit most popular brands of pipettes
  • Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
  • Tip sizes range from 10 to 1000 µl
  • Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
  • Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
  • Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates. View tip compatibility guides online at labcon.com/tipfits.
Certifications:
PFAS-free, Endotoxin-free, ATP-free, RNase/DNase-free, Low Trace Metals (<1 ppm), Protease-free, DNA-free.