Rival Low Extractable Powder-Free Nitrile Examination Gloves
Supplier: Techniglove
CP400 CP401 CP402 CP403 CP404 CP405
77888-586CS 405.27 CAD
77888-586 77888-592 77888-590 77888-588 77888-594 77888-596
Rival Low Extractable Powder-Free Nitrile Examination Gloves
Gloves
Rival 9.5" nitrile gloves are accelerator-free, powder-free, and low extractable. Rival’s new soft modulus nitrile delivers the fit and feel of latex for the ultimate stretch and comfort. Microtextured fingertips provide excellent grip on all surfaces.
  • Ambidextrous, powder-free, and latex-free
  • Textured fingertips
  • Accelerator-free
  • Force at break (N): >6
Rival gloves are 100% nitrile, contain no natural rubber, and are puncture-resistant. Environmentally and economically conscious gloves are packaged at 200 gloves per box to save space and reduce
More Product Information