Eclipse™ Pipette Tips for Rainin LTS Pipettes
Supplier: Labcon
1011-260-000-9 1011-960-008-9 1011-965-008-9 1018-260-000-9 1018-960-008-9 1018-965-008-9 1019-260-000-9 1019-960-008-9 1019-965-008-9
77871-134CS 558.53 CAD
77871-134 77871-136 77871-138 77871-140 77871-142 77871-144 77871-146 77871-148 77871-150
Eclipse™ Pipette Tips for Rainin LTS Pipettes
Pipette Tips
Labcon tips for LTS pipettes are made from special flexible resins. For easiest ejection apply less than 3 kg of force when mounting. Do not force tips onto the pipette barrel with excessive force.
  • Engineered to fit old and new Rainin LTS pipettes
  • Ergonomic, flexible top eliminates excessive mounting force
  • Ultrafine point ensures precision and accuracy in sample delivery
  • Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
  • Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
  • Made in USA
Labcon resins are pre-tested to ensure no extractables are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates.
Certifications:
All tips are Endotoxin-free, ATP-free, RNase/Dnase-free, Low Trace Metals (<1 ppm). Select tips are Protease-free, DNA-free, PFAS-free.