Cleanroom Gloves Dispensers
Supplier: PALBAM 2008 AC LTD TE
GDS-0605 GDS-6860 GDS-8760
77898-246EA 2210.74 CAD
77898-246 77898-282 77898-270
Cleanroom Gloves Dispensers
PPE Dispensers
Palbam Class is a world-renowned Cleanroom Gowning Design and Furniture supplier. The extensive range of gownroom products designed and manufactured by Palbam Class is second to none.
  • Designed for dispensing packaged sterile gloves
  • A slot the vertical length of each chamber and a top lid, allows easy refilling of gloves
  • Wall mounted
  • Electropolished 304 stainless steel
The Gownroom is often the first impression for employees and visitors alike, that they are entering into a special area. Palbam Class products are guaranteed to ensure that your company will give the best possible impression, as well as serving all the ergonomic, cleanliness and functional requirements.