ZAP™ Aerosol Filter Pipette Tips
Supplier: Labcon
1155-965-008-9 1051-225-000-9 1051-965-038-9 1055-225-000-9 1055-965-018-9 1057-225-000-9 1057-965-018-9 1059-225-000-9 1059-965-008-9 1153-225-000-9 1153-965-008-9 1154-225-000-9 1154-965-008-9 1155-225-000-9 1094-225-000-9 1094-965-008-9 1191-965-008-9
490008-004CS 794.6 CAD
490008-004 77871-174 77871-176 77871-178 77871-180 77871-182 77871-184 77871-186 77871-188 77871-236 77871-238 77871-240 77871-242 77871-244 77871-220 77871-222 77871-284
ZAP™ Aerosol Filter Pipette Tips
Pipette Tips
Labcon's ZAP™ aerosol filter pipette tips have been proven with case studies to block aerosol contamination. Unlike some leading brands of self-sealing aerosol-resistant pipette tips, ZAP™ tips have no cellulose gum, toluene solvent in the filter to mix with and contaminate the sample. ZAP™ tips will not wick fluids like cellulose-filled aerosol-resistant tips.
  • Patented 10 µm, hydrophobic filter blocks aerosols, preventing cross-contamination
  • Uniquely engineered to fit most popular brands of pipettes
  • Versatile design for various lab applications, featuring bevel point, extended, graduated, ultrafine tips, and more
  • Tip sizes range from 10 to 1000 µl
  • Pure USP Class VI, naturally hydrophobic resins provide high-quality tip uniformity
  • Sterile tips guarantee the highest sterility assurance level of 10⁻⁶
  • Made in USA
Labcon resins are pre-tested to ensure no extractables, PFAS, or other compounds are present.
All tips undergo continuous lot testing to ensure precision and consistent performance in Labcon in-house physical and biological testing labs. Download certificates by lot number at labcon.com/certificates. View tip compatibility guides online at labcon.com/tipfits.
Certifications:
PFAS-free, Endotoxin-free, ATP-free, RNase/DNase-free, Low Trace Metals (<1 ppm), Protease-free, DNA-free.