Corepoint® Scientific Stainless Steel Freezers, 25 cu. ft.
Supplier: Horizon Scientific
CPS-HC-SSF-25 CPS-HC-SSF-2530 CPS-HC-SSF-2530-LH CPS-HC-SSF-25-LH
77919-546EA 10402.14 CAD
77919-546 77919-548 77919-550 77919-552
Corepoint® Scientific Stainless Steel Freezers, 25 cu. ft.
Freezers Laboratory / General Purpose Freezers
These 25 cu. ft. freezers are built to last with their durable stainless-steel construction and energy-efficient performance.
  • Superior temperature uniformity
  • Microprocessor temperature controller
  • Audible and visual high and low temperature alarms
  • Probe access port
  • Natural hydrocarbon refrigerants
Designed for laboratories and medical facilities, these units deliver the accuracy and precision you need to support your critical substances. Experience exceptional microprocessor temperature control and enjoy environmentally-friendly refrigerants for energy
More Product Information