Vikan® UST Tank Brushes
Supplier: Remco Products
70301 70302 70303 70304 70305 70306 70307 70308
77906-514EA 137.14 CAD
77906-514 77906-508 77906-502 77906-520 77906-510 77906-516 77906-492 77906-498
Vikan® UST Tank Brushes
Brushes
Designed to incorporate Vikan®’s Ultra Safe Technology (UST), this industrial tank brush offers unparalleled bristle security and hygiene.
  • Angled bristles facilitate optimal cleaning
  • Wide drainage holes prevent water pooling and aid drying during storage
  • Threaded connection
  • Overall dimensions: 6.7W×4.1H×7.5"L
This UST tank scrub offers optimal performance in scrubbing efficacy, durability, and hygienic design. The seamless, easy-to-clean design minimizes the risk of contamination, making it an ideal tool for tank cleaning in dairies, breweries, wineries, and other food and beverage production facilities. The UST tank brush is compact and lightweight, making it perfect for cleaning the insides of complex equipment like mixers and blenders. A perfect fit for any color-coded cleaning system.
Certifications:
FDA compliant.