Cleanroom Sticky Mat Frames, Aluminum
Supplier: High-Tech Conversions
TTF-AL-1836 TTF-AL-1845 TTF-AL-2436 TTF-AL-2645 TTF-AL-3636 TTF-AL-3645 TTF-AL-3660
77898-372EA 537.04 CAD
77898-372 77898-376 77898-374 77898-382 77898-378 77898-370 77898-380
Cleanroom Sticky Mat Frames, Aluminum
Floor Mats Mat Frames
This cleanroom sticky mat frame provides extra protection, and a smooth edge for walking and moving wheeled carts over the mat.
  • Light weight tacky mat frame
  • Durable low profile design
  • Easy to clean
  • Non-skid backing
  • Aluminum material
Aluminum tacky mat frame features a sloping 4" wide anodized aluminum edge, non-skid backing, permanently secured plastic base, and measures 5/16" height. This mat frame is designed for cleanroom sticky mats and is available in the following sizes: 18×36", 24×36", 26×45", 36×36", 36×45", 36×60".
Ordering information:
Frame only. Cleanroom sticky mats are sold separately. Contact your Avantor representative for more information.