PIP® 4-in-1 AR/FR Dual Certified Ripstop Jacket with Zip-Out Liner
Supplier: PIP
385-FRFC-NV-2X 385-FRFC-NV-3X 385-FRFC-NV-4X 385-FRFC-NV-5X 385-FRFC-NV-L 385-FRFC-NV-M 385-FRFC-NV-S 385-FRFC-NV-XL
77898-412EA 544.15 CAD
77898-412 77898-434 77898-422 77898-436 77898-430 77898-424 77898-406 77898-428
PIP® 4-in-1 AR/FR Dual Certified Ripstop Jacket with Zip-Out Liner
Workwear
4-in-1 AR/FR ripstop jackets feature 8 pockets (shell: 2 chest patch, 1 sleeve pen, 2 waist patch, 1 interior; Liner: 2 waist slash, 1 sleeve pen) and an FR liner that can be worn alone or zipper into the outer shell.
  • Shell: 7 oz. Ripstop 88% cotton/ 12% nylon fabric with 7 oz. khaki FR twill weave fabric for lining
  • Liner: 7 oz. FR Ripstop 88% cotton/12% nylon fabric with 14 oz. khaki 100% Modacrylic lining
  • High tenacity Aramid FR thread, bar-tacked at all stress points
  • Protective stand-up collar with hook and loop closure
  • Detachable hood with 3 snaps that attach to jacket
  • Heavy-duty brass zipper, with storm guard and hook and loop closure
  • ATPV 26 Cal/cm2, CAT 3
  • Liner features detachable sleeves that convert into vest
The PIP® FR garments are engineered with high-performance materials to provide advanced thermal protection, flame resistance, and protection against arc flash exposure. Ideal for mining crews, petrochemical, oil and gas workers, utilities, and welding in cold climates.
Renewably sourced, non-fluorinated water repellent treatment. Overalls are reusable and launderable.
Certifications:
UL certified to NFPA 2112-2023, NFPA 70E, ASTM F1506, OEKO-TEX standard 100.
Packaging:
Individually bagged.