PIP® AR/FR Dual Certified Insulated Bib Overalls
Supplier: PIP
385-FRBO-NV-2X 385-FRBO-NV-3X 385-FRBO-NV-4X 385-FRBO-NV-5X 385-FRBO-NV-L 385-FRBO-NV-M 385-FRBO-NV-S 385-FRBO-NV-XL
77898-420EA 398.93 CAD
77898-420 77898-418 77898-432 77898-414 77898-426 77898-410 77898-416 77898-408
PIP® AR/FR Dual Certified Insulated Bib Overalls
Workwear
FR insulated bib overalls feature 6 pockets (2 chest patch, 2 slanted waist, 2 rear patch) and adjustable shoulder straps.
  • 7 oz. FR ripstop material, 88% cotton/ 12% nylon
  • 14 oz. 100% modacrylic insulation
  • ATPV 33 Cal/cm2/ Cat 3
  • Two-way, heavy-duty brass zipper on aramid tape at center front
  • 28" zipper at leg with storm guard and snap closure
  • Elasticized shoulder strap with adjustable buckles
The PIP® FR garments are engineered with high-performance materials to provide advanced thermal protection, flame resistance, and protection against arc flash exposure. Ideal for mining crews, petrochemical, oil and gas workers, utilities, and welding in cold climates.
Renewably sourced, non-fluorinated water repellent treatment. Overalls are reusable and launderable.
Certifications:
UL certified to NFPA 2112-2023, NFPA 70E, ASTM F1506, OEKO-TEX standard 100.
Packaging:
Individually bagged.