AssureTouch™ Guard Micro Polychloroprene Sterile Surgical Gloves, Powder-Free
Supplier: Medicom
1144-5.5 1144-6.5 1144-6 1144-7.5 1144-7 1144-8.5 1144-8 1144-9
CA77889-122CS 412.31 CAD
CA77889-122 CA77889-088 CA77889-084 CA77889-114 CA77889-078 CA77889-086 CA77889-074 CA77889-104
AssureTouch™ Guard Micro Polychloroprene Sterile Surgical Gloves, Powder-Free
Gloves
Choosing quality, latex-free, sterile polychloroprene surgical gloves eliminates the risk to patients and staff with latex sensitivity without giving up comfort, protection, performance, or compliance.
  • Highly durable and puncture-resistant
  • Sterile, latex-free, and powder-free
  • Textured palm
Polychloroprene combines nitrile-like strength with latex-like comfort, without the risks of Type I allergies or latex sensitization. Ideal for procedures requiring tactile sensitivity. Micro thickness allows for double gloving.
Certifications:
Meets ASTM D3577-19
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Disposable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Material Polychloroprene
Finish Textured palm
Cuff style Beaded
Finger thickness 5.12 mil
Palm thickness 4.30 mil
Sterility Sterile
ASTM No. D3577-19