VWR® Hood, Beard, and Shroud Covers
Supplier: VWR International
77610-718 77610-714
77610-718CS 1518.59 CAD
77610-718 77610-714
VWR® Hood, Beard, and Shroud Covers
Disposable Garments
The ideal solution for comprehensive facial protection. These hoods are non sterile and offer outstanding coverage, making them a favored choice. Crafted from breathable, non-shedding materials, these hoods are designed for cleanroom environments due to their unique design.
  • Serged seam construction
  • SMS three-layer polypropylene material
  • ⅛" elastic face opening
  • 25 gsm
  • Non sterile
Designed from non-woven polypropylene, these hoods are specifically suitable for cleanrooms. Beyond their compatibility, they are known for their comfort, soft texture, and excellent breathability.
Hood, beard, and shroud covers are packaged in color-coded packaging for quick and easy size recognition. Large covers are supplied in green and XL covers are supplied in blue packaging.
All components are manufactured in the USA.
Packaging:
200 pieces per bag, (poly-lined carton), double-bagged. 200 each per case.