AssureTouch™ Defend Polychloroprene Sterile Surgical Gloves, Powder-Free
Supplier: Medicom
1146-5.5 1146-6.5 1146-6 1146-7.5 1146-7 1146-8.5 1146-8 1146-9
CA77889-098CS 475.39 CAD
CA77889-098 CA77889-108 CA77889-120 CA77889-100 CA77889-072 CA77889-080 CA77889-110 CA77889-128
AssureTouch™ Defend Polychloroprene Sterile Surgical Gloves, Powder-Free
Gloves
Choosing quality, latex-free, sterile polychloroprene surgical gloves eliminates the risk to patients and staff with latex sensitivity without giving up comfort, protection, performance, or compliance.
  • Highly durable and puncture-resistant
  • Sterile, latex-free, and powder-free
  • Textured palm
Polychloroprene combines nitrile-like strength with latex-like comfort, without the risks of Type I allergies or latex sensitization. Mint green color aids in instant identification as non-latex and quicker detection of tears when double gloving. Standard thickness makes them suitable for a wide range of surgical procedures.
Certifications:
Meets ASTM D3577-19.
Packaging:
Gloves are packaged 50 pairs per box, 4 boxes per case (200 pairs per case).