Sorenson™ LTS Compatible TipStation Pipette Tips
Supplier: Corning
88030 88040 88050 88060
77775-492CS 395.16 CAD
77775-492 77775-494 77775-496 77775-498
Sorenson™ LTS Compatible TipStation Pipette Tips
Pipette Tips
LTS compatible TipStation pipette tips are available sterile or non sterile in 20 and 200 µl, and are a perfect platform for multi-channel pipetting.
  • Compatible with LTS pipettors
  • Ideal platform for multi-channel pipettors or high throughput laboratories
  • Can position lid to expose 96 or 192 tips
  • Certified RNase/DNase free, human DNA free, and non-pyrogenic
Certifications:
ISO 13485, ISO 14001.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable