Perform sample transfers between virtually any tube, rack, microplate, or horizontal gel box quickly and efficiently with adjustable tip spacing.
Adjustable tip spacing allows users to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Advanced connectivity enables access to Thermo Fisher Scientific cloud-based pipetting apps. Tip is locked and sealed in place until ejected, so there are no dropped tips that waste valuable sample. Complete seal to secure the sample volume in each tip for enhanced accuracy and precision. Tips are attached and ejected with a light touch. This eliminates the frustration of banging tips on the pipette.
Customers can utilize my pipette creator, a new web-based pipetting app powered by Thermo Fisher cloud, to revolutionize pipetting with a new era of technology. This enables efficient, centralized programming and sharing of protocols between E1-ClipTip™ pipettes and colleagues. The app offers pre-programmed protocols, to download and share, for many of the favorite reagent kits and pipetting protocols. Connect E1-ClipTip™ pipette to the app via a Bluetooth or USB connection.
E1-ClipTip™ is designed for optimum comfort and ease of use. The electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting. Step-based Matrix or Presets with settings for the most common applications are two operating options available. The Programs function allows storage of 30 of the most common Matrix or Presets protocols with a specific name. Five different user profiles offer customized settings for multiple users and protocols. The MyPipette™ function allows customizing the main menu view by organizing the icons in a preferred order or creating shortcuts for favorite programs and pipetting functions. Protect important settings from unintentional editing with a password. Service and calibration trackers offer reminders when the pipette needs to be sent for service. Pipettors run on a Li-ion battery for long-lasting performance and fast recharging. Recharging can be done with a charging stand or a plug.
Available formats include Microcentrifuge tubes to/from 24,-48,-96-and 384- well microplates; Test tubes to/from deepwell blocks; Microplates and tubes to agarose gels. Save up to 90% in time and repetitions compared to use of single channel manual pipettes.
Accessories information: A wide selection of ClipTip™ tips are available. ClipTip™ 96-format tips are available in 20, 50, 200, 300, 1000, and 1250 µl. ClipTip™ 384-format tips are available in 12.5, 30, and 125 µl. Tips come in filtered, sterile, non-sterile, racked, and reload packaging options.
Ordering information: Pipettors come with a 2-year manufacturer's warranty with convenient web registration.