Ghost-Buster Columns
Supplier: WELCH MATERIALS, INC.
06100-31001 GBKIT-02 GBKIT-03 GBKIT-04 06100-31008 06100-31016 06100-31018 06100-31021 06100-31025 06100-31026 06100-31027 GBKIT-01
77865-738EA 1226.32 CAD
77865-738 77865-756 77865-758 77865-760 77865-740 77865-742 77865-744 77865-746 77865-748 77865-750 77865-752 77865-754
Ghost-Buster Columns
Chromatography Columns
The Welch ghost-buster column eliminates ghost peaks in chromatographic separation by removing low-polarity impurities from the mobile phase, mixer, and pipelines. Installed between the gradient mixer and injector, it adsorbs weak-polar and non-polar organic contaminants without altering the mobile phase composition. Unlike in-line filters, it targets organic pollutants rather than just solid particles. This improves separation clarity, prevents interference, and extends the lifespan of both the column and the system, making it ideal for long-term, high-precision operations.
  • Efficiently eliminates ghost peaks and baseline fluctuation
  • Removes impurities from the mobile phase
  • Enables reliable and robust analysis of trace compounds
Welch ghost-buster columns can effectively remove impurities with low polarity and thus prevent the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well.
Certifications:
ISO 9001 quality standard.
Caution:
These columns can not be opened or repacked.