Vertex® Pipette Tips and EcoPac® Reloads
Supplier: SCIENTIFIC SPECIALTIES, INC.
4115N00S 4117N00S 4117NS0S 4135N00S 4137N00S 4137NS0S 4235N00S 4237N00S 4237NS0S 4335N00S 4337N00S 4337NS0S 4345N00S 4347N00S 4347NS0S 4925N00S 4937N00S 4937NS0S
77871-098CS 506.14 CAD
77871-098 77871-100 77871-102 77871-104 77871-106 77871-108 77871-110 77871-112 77871-114 77871-116 77871-118 77871-120 77871-122 77871-124 77871-126 77871-128 77871-130 77871-132
Vertex® Pipette Tips and EcoPac® Reloads
Pipette Tips
Clean liquid release for maximum sample recovery and increased accuracy. Vertex Pipette Tips feature our proprietary NoStick® resin that provides researchers with exceptional liquid handling control. Low-retention properties facilitate a much cleaner release, dispensing all of the sample and improving pipetting accuracy.
  • Universal pipettor compatibility
  • High-clarify virgin polypropylene (meets FDA 21 CFR 177.1520 and USP Class VI)
  • Unique NoStick® resin technology facilitates clean release
  • Certified free of detectable RNase, DNase, DNA, PCR inhibitors and tested pyrogen free
Vertex® EcoPac® Refill systems provide handy lift-and-load simplicity, and do not require any tools or special instructions to move from an empty insert to a fresh set of 96 tips. EcoPac® Refill systems are also environmentally friendly, reducing lab waste by 70% and requiring less shelf space than standard racks.
Made in the USA.
Ordering information:
For Vertex® filter tips, see series 76808-776.