PROCLEAR Polypropylene DEMI Capsules
Supplier: PARKER HANNIFIN TE
PEPPE-01GG-PS3-FB PEPPE-05GG-PS3-FB PEPPA-01GG-PN3 PEPPA-01GG-PS3 PEPPA-01HH-PN3 PEPPA-01HH-PS3 PEPPA-03GG-PN3 PEPPA-03GG-PS3 PEPPA-03HH-PN3 PEPPA-03HH-PS3 PEPPA-05GG-PN3 PEPPA-05GG-PS3 PEPPE-01GG-PN3 PEPPE-01GG-PS3 PEPPE-01HH-PN3 PEPPE-01HH-PS3 PEPPE-03GG-PN3 PEPPE-03GG-PS3 PEPPE-03HH-PN3 PEPPE-03HH-PS3 PEPPE-05GG-PN3 PEPPE-05GG-PS3 PEPPB-01GG-PN3 PEPPB-01GG-PS3 PEPPB-01HH-PN3 PEPPB-01HH-PS3 PEPPB-03GG-PN3 PEPPB-03GG-PS3 PEPPB-03HH-PN3 PEPPB-03HH-PS3 PEPPB-05GG-PN3 PEPPB-05GG-PS3 PEPPB-05HH-PN3 PEPPB-05HH-PS3 PEPPB-10GG-PN3 PEPPB-10GG-PS3 PEPPB-10HH-PN3 PEPPB-10HH-PS3 PEPPB-96GG-PN3 PEPPB-96GG-PS3 PEPPB-96HH-PN3 PEPPB-96HH-PS3 PEPPA-05HH-PN3 PEPPA-05HH-PS3 PEPPA-10GG-PN3 PEPPA-10GG-PS3 PEPPA-10HH-PN3 PEPPA-10HH-PS3 PEPPA-96GG-PN3 PEPPA-96GG-PS3 PEPPA-96HH-PN3 PEPPA-96HH-PS3 PEPPE-05HH-PN3 PEPPE-05HH-PS3 PEPPE-10GG-PN3 PEPPE-10GG-PS3 PEPPE-10HH-PN3 PEPPE-10HH-PS3 PEPPE-96GG-PN3 PEPPE-96GG-PS3 PEPPE-96HH-PN3 PEPPE-96HH-PS3 PEPPB-01GG-PS3-FB PEPPE-05GG-PN3-FB
76554-134PK 342.32 CAD
76554-134 76554-136 77296-340 77296-342 77296-344 77296-346 77296-348 77296-350 77296-352 77296-354 77296-356 77296-358 77296-420 77296-422 77296-424 77296-426 77296-428 77296-430 77296-432 77296-434 77296-436 77296-438 77296-380 77296-382 77296-384 77296-386 77296-388 77296-390 77296-392 77296-394 77296-396 77296-398 77296-400 77296-402 77296-404 77296-406 77296-408 77296-410 77296-412 77296-414 77296-416 77296-418 77296-360 77296-362 77296-364 77296-366 77296-368 77296-370 77296-372 77296-374 77296-376 77296-378 77296-440 77296-442 77296-444 77296-446 77296-448 77296-450 77296-452 77296-454 77296-456 77296-458 77437-870 77437-872
PROCLEAR Polypropylene DEMI Capsules
Filters Membrane Filter Capsules
PROCLEAR polypropylene filters are designed for a wide range of prefiltration duties within the production of pharmaceuticals. They are particularly suited for applications where chemical compatibility is an issue.
  • Graded density polypropylene media for high capacity
  • Extremely robust to withstand aggressive conditions
  • All polypropylene construction
  • MURUS and DEMICAPs can be gamma-irradiated and autoclaved
  • PFAS-free options available
The optimum pleat configuration and graded density polypropylene media used in PROCLEAR PP filters ensure the filters have the highest possible throughput to blockage, resulting in long service life.
The PROCLEAR PP range of filters are fully supported by a comprehensive validation guide to simplify its specification into new and existing processes.
Additional connection configurations are available.
Packaging:
DEMI Capsules are supplied three units per pack. FB Extension stand for filling bell included.