Saturix Disposable Heavy-Weight Microfiber Mops
Supplier: FG CLEAN WIPES TE
MD416 MD416S
77871-068CS 789.21 CAD
77871-068 77871-070
Saturix Disposable Heavy-Weight Microfiber Mops
Mops
Optimized cleaning performance and efficiency. Mops are available in non sterile or validated sterile formats.
  • Removes 99.9% of particles down to 0.1 µm in size
  • 28 m²/ 305 sq. ft. coverage per mop, 47% more than leading competitors
  • Pocket mop design facilitates hands-free operation and minimizes the spread of dirty cleaning solutions
  • High absorption and coverage to maximize worker efficiency
  • Consistent and even laydown of cleaning solutions
  • Certificate of sterility included with sterile mops
  • 3-year shelf life
Microfiber cleaning power coupled with the hands-free mop drop system provides consistent cleanliness and minimizes the spread of dirty cleaning solutions while improving productivity. The exacting sterilization process offers absolute confidence for use with sterile mops.
Packaging:
Mops are packaged 5 per bag, 20 bags per case.