Saturix Stainless Steel Frame for Pocket Mops, Mop Drop Compatible
Supplier: FG CLEAN WIPES TE
HF216 HH270
77871-064EA 499.77 CAD
77871-064 77871-066
Saturix Stainless Steel Frame for Pocket Mops, Mop Drop Compatible
The highest performing Saturix mop frame available.
  • Corrosion-resistant and durable premium V4A stainless steel
  • Fully autoclavable with at least 50 cycles
  • Top and bottom ridges for complete mop surface contact
  • Angle lock for effective ceiling and high surfaces cleaning
  • Quick connect for ease of use
  • Hands-free operation improves efficiency while minimizing the spread of dirty cleaning solutions
  • Seamless integration with Saturix cleaning carts
Fully autoclavable with premium materials, every detail of this stainless steel mop frame is designed to improve outcomes and efficiency.