NanoFil™ Gas-Tight Injection Systems
Supplier: WORLD PRECISION INSTRUMENTS LLC
NANOFIL NANOFIL-100
77664-898EA 221.2 CAD
77664-898 77664-900
NanoFil™ Gas-Tight Injection Systems
Syringes Syringes, Medical
The NanoFil™ Gas-Tight Injection System boasts zero dead volume. Variability is eliminated with the NanoFil™  design: our interchangeable needles advance directly into the syringe barrel, creating a 1:1 connection with the plunger-a truly gas-tight system built for low-volume sample control.
  • Virtually zero dead volume microinjection syringe
  • Strong gas-tight seal providing both repeatability and superior application outcomes
  • NanoFil™ needle sizes offered from 26G, down to 36G
  • Two needle types available-blunt and bevelled
  • Versatile for life science research applications requiring precise in vivo targeting, low-volume fluid transfer
  • Compatible with WPI’s UMP3 microinjection system
NanoFil™ promotes a range of applications, supported by our universal NanoFil™ needles - where all gauges are fully interchangeable and compatible with all NanoFilTM syringes. NanoFil™ Intraocular (IO) and Retinal Pigment Epithelium (RPE) kits diversify the integration for fine-tuned fluid transfer into sensitive tissue, mitigating user error. NanoFil™ syringes easily attach to WPI’s UMP3 syringe pump paired to a stereotaxic or manipulator-fixed setups for enhanced control.
Gas-tight control is particularly critical for applications requiring utmost accuracy and precision during low-volume fluid handling. Having a quality gas-tight design ensures a vacuum-like seal between the syringe and needle, supplying the necessary force to advance small samples forward during delivery, and upwards during withdraw.
NanoFil’s virtual zero dead-volume comes from its thoughtfully designed coupling mechanism, where all WPI NanoFil™ needles insert directly into the syringe barrel, creating a 1:1 connection with the syringe’s plunger. Priming your syringe is made effortless with unparalleled control, supported by a sure-seal, gas-tight system.
NanoFil™ syringes are packaged with a bevelled 26G needle, but smaller gauges are usually preferred for in vivo applications requiring localized targeting of sensitive soft tissues, as well as tedious ex-vivo low-volume sample dispensing. NanoFil™ needles come in 26, 33, 34, 35, and 36G blunt or bevelled styles. Each needle is handcrafted and individually tested by the WPI team to ensure quality and functionality. Currently, WPI carries the smallest commercially available needle gauge on the market at 36G. The sharpness of our unique tri-tip bevel offers a less invasive, smooth entry into tissue compared to single-cut bevels offered by competitors.
Features and applications:
Biologically compatible injection syringe
Compatible with WPI's UMP3 microinjector for precise, targeted injections
Animal research, viral targeting (e.g. AAV construct, DREADDS): Retinal Pigment Epithelial (RPE)/ Intra Ocular (IO), Brain, Muscle, Spine
Capillary electrophoresis