Cryo Gloves and Protective Wear, Thermo Fisher Scientific
Supplier: Thermo Fisher Scientific
4425 4426
89176-742EA 732.63 CAD
89176-742 89176-744
Cryo Gloves and Protective Wear, Thermo Fisher Scientific
Gloves
Keep your hands protected from ultra-low and cryo temperatures with these highly insulated cryo gloves.
  • Comfortable, lightweight, and breathable protection
  • Available in a variety of sizes
Certifications:
CE
Packaging:
One pair of cryo gloves provided.