Micro Nitric Oxide Sensors, World Precision Instruments
Supplier: WORLD PRECISION INSTRUMENTS LLC
ISO-NOP007 ISO-NOP3005 ISO-NOP3020
77664-476PK 1772.76 CAD
77664-476 77664-478 77664-480
Micro Nitric Oxide Sensors, World Precision Instruments
Electrodes Dissolved Oxygen Electrodes
The ISO NOP007 has a tip diameter of just 7 µm and a length of 2 mm. The response is linear over a wide dynamic concentration range of NO.
  • Excellent selectivity to NO
  • Rapid response time
  • Highly sensitive
  • Response time: <3 seconds
  • Poise voltage: 865 mV
  • Linear response over a large dynamic concentration range of NO
  • Dry electrodes require no sleeves or filling solution
The design is based on a single carbon fiber coated with WPI's no-selective membrane. A detection limit of approximately 1 nM no makes this electrode ideal for use in tissues and microvessels.
The ISO NOP30 has a tip diameter of 30 microns and is available in two different tip lengths ISO NOP3020 has tip length of 2 mm, ISO-NO3005 has tip length of 0.5 mm). The response is linear over a wide dynamic concentration range of no. The design is based on a single carbon fiber coated with WPI's no-selective membrane. A detection limit of approximately 1 nM no makes these electrodes ideal for use in tissues and microvessels.
Ordering information:
Requires cable (77663-980) sold separate.
Packaging:
Package of 3.