Pre-Pulled Glass Pipettes, Long Taper
Supplier: WORLD PRECISION INSTRUMENTS LLC
TIP10FLT TIP10LT TIP15FLT TIP15LT
77665-246EA 172.22 CAD
77665-246 77665-248 77665-260 77665-262
Pre-Pulled Glass Pipettes, Long Taper
Pipettes Serological Pipettes
Pre-pulled glass pipettes save the cost and time of making your own micropipettes. Each micropipette is pre-pulled for microinjection and are individually vacuum packed.
  • Pulled glass
  • Long taper
  • Pre-pulled tip for microinjection that requires depth
  • Every pipette individually vacuum packed
  • Glass outside diameter: 1.0 or 1.5 mm
  • 10 per package