VWR® Premium Series Upright Ultra-Low Temperature Freezers, 230 V / 50 Hz
Supplier: VWR International
77618-806 77618-770 77618-782 77618-794
77618-806EA 29928.36 CAD
77618-806 77618-770 77618-782 77618-794
VWR® Premium Series Upright Ultra-Low Temperature Freezers, 230 V / 50 Hz
Freezers Ultra Low Temperature Freezers
The VWR® Premium Series ULT freezers provide the power of performance with energy efficiency for a greener tomorrow.
  • 230 V / 50 Hz models
  • Operate between -50 to -86 °C
  • 4 storage capacities: (400, 500, 600, 700 2" cryoboxes)
  • ENERGY STAR® certified
  • Easy-to-use touch button interface
  • Vacuum insulation panels and high density water-blown foam
  • USB for easy export of recorded temperatures and events
  • 5 year warranty parts and labor
Experience the power of these premium ULT freezers, meticulously engineered to excel in the most demanding laboratory settings. With precise temperature control, minimal peak variation, and fast pull-down and recovery times, these freezers consistently deliver the perfect conditions your critical samples demand.
Sustainability meets savings with the Energy Star-certified VWR ULT premium freezers. Designed to consume less energy without sacrificing performance, these freezers help reduce operational costs and environmental impact. Our ULT freezers empower your laboratory to achieve its sustainability goals and promote a greener future.
The VWR premium ULT freezers are built for seamless operation with user-friendly features at its core. These whisper-quiet freezers provide an intuitive interface and self-serve updates to help minimize downtime. With a relocated USB port for quicker data retrieval, flexible rack configurations and faster re-entry times, the premium ULT freezers are designed to enhance the overall user experience and meet the needs of busy laboratories.
Certifications:
ENERGY STAR, cULus, CE
Ordering information:
Accessories and racks must be ordered separately.
Caution:
Must be removed from pallet upon delivery. Fits through standard 34.5" doorways.
230 V models. Corresponding models with voltage suitable for use in US (77618-762, 77618-774, 77618-786 and 77618-798) are also available, please see separate product page.