KITE Manual Micromanipulator
Supplier: WORLD PRECISION INSTRUMENTS LLC
KITE-M3-L KITE-M3-R
77664-510EA 1738 CAD
77664-510 77664-512
KITE Manual Micromanipulator
Micromanipulators
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 tilting base. Our 5 lbs. weight is frequently sold with this unit to provide stability.
  • Control knobs clustered in the same plane for quick resolution
  • Right and left hand orientation options available
Applications:
Microinjection
Electrophysiology recording
Mounting the KITE manipulator:
When the KITE is mounted on an M-3 tilting base with the steel weight and tilted at 45°, the average maximum practical height from the tabletop to the center of the pivot point on the manipulator is approximately 20 cm.
Economy manual manipulator:
Vernier scales allow readings to 0.1 mm.
X-axis fine control allows readings to 10 µm.
Choice of optional M-3  Tilting base which can be mounted to a table with M6 screws. 5-lb weight for tilting base sold separately.
Left or right-handed versions of the KITE micromanipulator are supplied with a standard 12 mm clamp and electrode holder.