MicroFil Flexible Needle
Supplier: WORLD PRECISION INSTRUMENTS LLC
MF28G-5 MF28G67-5 MF34G-5
77664-810EA 132.72 CAD
77664-810 77664-812 77664-814
MicroFil Flexible Needle
Syringe Needles
MicroFil fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles.
The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil plastic needle may be stored for days with the filling solution inside without clogging.
Tip Flexibiliy:
The MicroFil's tip elasticity is sturdy and very flexible though not unbreakable. Since it is more flexible than stainless steel needles, moderate bending will not block or damage the MicroFil needle. The combination of plastic and fused silica in the MicroFil tip is sturdier than plastic tips, allowing easy and repeated insertions into micropipettes. MicroFil's luer fitting allows easy coupling to syringes and syringe filters.
Autoclavability:
HDPP High Density Polypropylene–Autoclavable
Polyimide coating–Autoclavable