DuPont™ Tychem® 6000 SFR Coveralls
Supplier: DuPont
FS127TLT2X000600 FS127TLT3X000600 FS127TLT4X000600 FS127TLT5X000600 FS127TLTLG000600 FS127TLTMD000600 FS127TLTSM000600 FS127TLTXL000600
77889-276CS 931.81 CAD
77889-276 77889-274 77889-280 77889-268 77889-278 77889-270 77889-282 77889-272
DuPont™ Tychem® 6000 SFR Coveralls
Disposable Garments
DuPont™ Tychem® 6000 SFR coveralls feature a respirator fit hood, front zipper closure, chin flap, double storm flap, elastic face opening, elastic wrists, elastic ankles, elastic waist, taped seams, and pin lock zipper.
  • Respirator-fit hood
  • Covered elastic face opening
  • Covered elastic openings at wrists and ankles for tighter fit
  • Elastic at sides of waist for better fit
  • Chin flap with double-sided adhesive tape
  • Double storm flap covers the zipper, which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion
  • Taped seams provide strong chemical resistance against heavy liquid splashes
A sewn seam is heat-sealed with a strip of compatible chemical-resistant material, featuring a nylon zipper with a large metal pull and a pin lock mechanism that prevents accidental unzipping, all manufactured without natural rubber latex.
DuPont™ Tychem® 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist. Designed to be worn over primary flame-resistant protective clothing, Tychem® 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires. Tychem® 6000 SFR fabric provides at least 30 minutes of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
In the event that a flash fire occurs, Tychem® 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment, such as DuPont™ Nomex®, is worn beneath.
Applications include use in oil and gas industries, industrial chemical processing plants, laboratories, hazmat teams in fire departments, industrial fire brigades, and clandestine lab investigation.
Certifications:
Coveralls meet the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671), protecting against several bloodborne pathogen exposure risks.