Velocity™ Compact Cable Lockout Padlocks, Brady
Supplier: Brady Worldwide
177447 177448 177449 177450 177451 177452 177453 177464 177465 177466 177467 177468 177469 177470 177471 177472 177454 177455 177456 177457 177458 177459 177460 177461 177462 177463
77873-694EA 44.05 CAD
77873-694 77873-734 77873-698 77873-770 77873-790 77873-774 77873-764 77873-732 77873-780 77873-768 77873-792 77873-740 77873-720 77873-760 77873-726 77873-778 77873-702 77873-746 77873-786 77873-794 77873-714 77873-772 77873-700 77873-710 77873-686 77873-784
Velocity™ Compact Cable Lockout Padlocks, Brady
Security Locks
Velocity™ Compact cable lockout padlocks are engineered smaller and more versatile for complex or unique lockout applications.
  • Service temperature: −15 to +250 °F
  • Body size: 1.25W×0.625D×1.25" H
Compact padlock body with a flexible yet durable cable shackle lets users secure multiple or hard-to-reach isolation points. Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable, and exclusively for safety. Color-matched keys and padlock bodies help workers find the right key for the right lock fast. Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities.
Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection. Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access. Key codes engraved on the key and lock body to simplify record-keeping and traceability.
Certifications: OSHA 1910.147.
Ordering information:
Individual (each) padlock orders include 1 key, 1 padlock, 1 English label, 1 French label, and 1 Spanish label. Padlocks ordered in a pack of 6 include 6 keys, 6 padlocks, 6 English labels, 6 French labels, and 6 Spanish labels.