Human Recombinant FLT-3 Ligand (from E. coli)
Supplier: VWR International
77751-114
77751-110
77751-112
77751-126
77751-114EA
9088.08
CAD
77751-114
77751-110
77751-112
77751-126
Human Recombinant FLT-3 Ligand (from E. coli)
Proteins and Peptides
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signalling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3). Human FLT-3 ligand is active on mouse cells.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.