Human Recombinant LEPTIN (from E. coli)
Supplier: VWR International
77751-414 77751-416 77751-418 77751-420
77751-414EA 1101.1 CAD
77751-414 77751-416 77751-418 77751-420
Human Recombinant LEPTIN (from E. coli)
Proteins and Peptides
Leptin is a hormone that is produced by adipose tissue and plays critical roles in the physiologic regulation of body weight. Leptin acts through the leptin receptor (LEPR) to regulate adipose mass by inhibiting hunger and balancing energy usage. Leptin mutations cause severe hereditary obesity and hypogonadism in rodents and humans. Leptin also has thermogenic actions, regulates enzymes of fatty acid oxidation, and is involved in hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.