Human Recombinant FGF-4 (from E. coli)
Supplier: VWR International
VWR100-31-1MG VWR100-31-25UG VWR100-31-500UG VWR100-31-5UG VWR100-31-100UG
77751-334EA 6814.07 CAD
77751-334 77751-336 77751-338 77751-340 77751-332
Human Recombinant FGF-4 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 4 (FGF-4) is a secreted growth factor that is predominantly expressed during bone morphogenesis and embryonic limb development.  FGF-4 is an important growth regulator for stem cells, fibroblasts, and endothelial cells.  FGF-4 contains a single N-linked glycosylation signal.  In-vitro studies suggest that unglycosylated FGF-4 is cleaved into 13 kDa and 15 kDa truncated proteins that have greater biological activity than the wild type 19 kDa FGF-4 protein.  Human FGF-4 shares high homology and is cross-reactive with mouse FGF-4. 
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.