CytoFLEX nano Flow Cytometers
Supplier: Beckman Coulter
D00710 D02028
CA77874-706EA 301459 CAD
CA77874-706 CA77874-710
CytoFLEX nano Flow Cytometers
Cell Counters and Flow Cytometers Flow Cytometers
The CytoFLEX nano Flow Cytometer is the newest member of the CytoFLEX platform.
  • Specifically designed for the analysis of nanoparticles
  • CytoFLEX nano Flow Cytometer overcomes the limitations of traditional flow cytometry
  • Provides researchers a sensitive, consistent, and flexible solution to advance nanoparticle research
The CytoFLEX nano Flow Cytometer is a pivotal development in flow cytometry that allows users to analyze extracellular vesicles (EVs) at least as small as 40 nm with ease. It simultaneously offers up to 6 separate fluorescent channels of detection and 5 side scatter channels to deliver full characterization and open additional avenues for nanoparticle research.
Accessories information:
CytoFLEX nano QC Scatterspheres are system reagents for quality control processes (144 nm polystyrene scattersphere), used to monitor scatter sensitivity in VSSC1, VSSC2, BSSC, YSSC, and RSSC channels on the CytoFLEX nano Flow Cytometer (3 bottles/pack). CytoFLEX nano QC Fluorospheres are system reagents for quality control processes (500 nm polystyrene beads), used to monitor fluorescence sensitivity in fluorescent detection channels on the CytoFLEX nano Flow Cytometer (3 bottles/pack). CytoFLEX nano Multi-intensity Fluorospheres are system reagents (500 nm polystyrene beads) for use with the Sensitivity Monitor feature in CytExpert nano software for the CytoFLEX nano Flow Cytometer (1 bottles/pack). CytoFLEX Cleaner is an EU REACH-compliant, enzymatic azide-free cleaning solution (5 L).