Human Recombinant PTN (from E. coli)
Supplier: VWR International
77751-602 77751-604 77751-606 77751-608 77751-610
77751-602EA 1049.25 CAD
77751-602 77751-604 77751-606 77751-608 77751-610
Human Recombinant PTN (from E. coli)
Proteins and Peptides
Pleiotrophin (PTN) is a heparin-binding growth factor that has mitogenic effects on fibroblast, epithelial, and endothelial cells. PTN is made by many tissues, but is predominantly secreted by nervous tissue during development. PTN induces neurite outgrowth and is involved in tumor growth and metastasis. PTN binds with low affinity to the cell surface receptor nucleolin to inhibit HIV-1 infection. PNT also binds the receptor protein tyrosine phosphatase type Z (PTPRZ), syndecan-3, and anaplastic lymphoma kinase (ALK) receptors.
  • High quality
  • Low endotoxin
  • Lot-to-lot consistency
  • Animal free
  • Manufactured following GMP guidelines
Caution: For research use or further manufacturing only.