Pig Recombinant IL-2 (from E. coli cells)
Supplier: VWR International
77753-288
77753-290
77753-292
77753-294
77753-296
77753-288EA
1807.25
CAD
77753-288
77753-290
77753-292
77753-294
77753-296
Pig Recombinant IL-2 (from E. coli cells)
Proteins and Peptides
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.