Rat Recombinant IGF-I (from E. coli cells)
Supplier: VWR International
77752-800
77752-802
77752-804
77752-806
77752-808
77752-800EA
686.19
CAD
77752-800
77752-802
77752-804
77752-806
77752-808
Rat Recombinant IGF-I (from E. coli cells)
Proteins and Peptides
Insulin-like growth factor I (IGF-I) is a growth factor that is produced by the liver. IGF-1 production is stimulated by growth hormone (GH). IGF-I binds the insulin-like growth factor 1 receptor (IGF1R) and the insulin receptor to stimulate systemic body growth. IGF-I is one of the most potent activators of the AKT signaling pathway, which stimulates cell proliferation and inhibits programmed cell death.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.