MinION™ Mk1D DNA/RNA Sequencing Device
Supplier: Oxford Nanopore Technologies
SUPP1D MDIP-RAD14 MDIP-RBK14.24 MDIP-RNA MDIP-RPB114 MDP-LSK114 MDP-NBD14.24 MDP-PCS14 MDP-RAD14 MDP-RBK14.24 MDP-RNA MDP-RPB114 BASIC12M-MK1D MDIP-LSK114 MDIP-NBD14.24 MDIP-PCS14
77867-966EA 9480 CAD
77867-966 77874-660 77874-662 77874-664 77874-666 77874-668 77874-670 77874-672 77874-674 77874-676 77874-678 77874-680 77874-642 77874-654 77874-656 77874-658
MinION™ Mk1D DNA/RNA Sequencing Device
Next Generation Sequencing Instruments
MinION™ - your personal, portable DNA and RNA sequencer.
  • Any length read from short to ultra long: 20 bp to >4 Mb
  • High-accuracy, direct sequencing of native DNA and RNA with built-in modification and best-in-class methylation detection
  • Real-time sequencing for immediate access to actionable results
  • Simple 10-minute sample prep available
  • Portable, pocket-sized device sequence in the field or lab
A sequencer for everyone - empowering individual researchers, labs and those new to DNA/RNA sequencing to perform in-house sequencing and take control of their timelines with a cost-effective, personal device. Weighing only 130 g and running off a laptop, MinION™ Mk1D generates tens of gigabases of real-time data in the field or lab.
The MinION™ Mk1D is a cutting-edge, portable sequencer enabling real-time sequencing in diverse environments. Designed to connect with modern laptops via its USB-C port, it is compatible with MinION™ Flow Cells.
Improved thermal regulation capabilities of the MinION™ MK1D significantly enhance sequencing performance, allowing highly accurate real-time sequencing in an even wider range of environments than its predecessor, the MinION™ Mk1B. Intuitive status LEDs make the status of the device clear at a glance.
The MinION™ Mk1D device is suitable for a range of applications, including metagenomic species identification, antibiotic resistance profiling, microbial genome assembly, plasmid sequencing, targeted sequencing, low-coverage sequencing of large genomes and isoform-level transcriptomics.
Ligation sequencing and native barcoding kits – highest output and control over read lengths (60 min. prep time; 1,000 ng gDNA).
Rapid sequencing and rapid barcoding kits – fastest, simplest library preparation (from 10 min. prep time; 200 ng gDNA). These kits are able to generate sequencing accuracies of over 99% (Q20+) and duplex data but have been optimized for speed and simplicity. If users require high duplex data, we recommend the ligation sequencing kit v14 or native barcoding kit v14.
Rapid PCR barcoding kit – ideal for low input amounts (15 min. + PCR prep time; 1 - 5 ng gDNA)
cDNA-PCR sequencing kit – identification and quantification of full-length transcripts with highest output (~225 min. + PCR prep time; 10 ng poly-A+ RNA or 500 ng total RNA).
Direct RNA sequencing kit – sequence RNA molecules directly and identify base modifications (135 min. prep time; 300 ng poly-A+ RNA or 1 μg total RNA).
Ordering information: For more information on nanopore sequencing library prep kits and flow cells, please visit the individual product pages. MinION™ rapid starter training is also available to ensure the successful implementation of nanopore sequencing. This is an online, two-day, personalized training course for up to two participants. Contact your Avantor representative to find out more and discuss your training needs.
Caution: These products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.