Human Recombinant GROWTH HORMONE (from E. coli)
Supplier: VWR International
77751-452 77751-454 77751-456 77751-458 77751-460
77751-452EA 151.6 CAD
77751-452 77751-454 77751-456 77751-458 77751-460
Human Recombinant GROWTH HORMONE (from E. coli)
Proteins and Peptides
Growth hormone (GH) is an important mitogenic growth factor that is synthesized, stored, and secreted by somatotropic cells of the anterior pituitary gland. GH stimulates growth, cell reproduction, and cell regeneration. In children, GH deficiencies can cause short stature, growth failure, and delayed sexual maturity.
  • High quality
  • Low endotoxin
  • Affordable
Adult GH deficiency presents with reduced lean body mass, increased adiposity, reduced muscle strength, and ultimately premature mortality. GH replacement therapy is used to treat many growth disorders, including Turner syndrome, chronic renal failure, and Prader-Willi syndrome.
Caution: For research use only.