Human Recombinant VEGF-121 (from E. coli)
Supplier: VWR International
77750-224
77750-226
77750-228
77750-224EA
1196.85
CAD
77750-224
77750-226
77750-228
Human Recombinant VEGF-121 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways.
VEGF-A has several alternatively spliced isoforms, with one being VEGF-121. The VEGF-121 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell function.