Human Recombinant VEGF-121 (from E. coli)
Supplier: VWR International
77750-216 77750-218 77750-220 77750-222
77750-216EA 1101.1 CAD
77750-216 77750-218 77750-220 77750-222
Human Recombinant VEGF-121 (from E. coli)
Proteins and Peptides
Vascular endothelial growth factor-A (VEGF-A) is produced by a wide variety of cell types, including tumor and vascular cells. VEGF-A is a mediator of vascular growth, vascular permeability, and plays a role in stimulating vasodilation via nitric oxide-dependent pathways.
  • High quality
  • Low endotoxin
  • Affordable
VEGF-A has several alternatively spliced isoforms, with one being VEGF-121. The VEGF-121 isoform is a secreted protein that acts on receptors VEGFR-1 and VEGFR-2 to modulate endothelial cell function.
Caution:
For research use only.