Human Recombinant EXODUS-2 (from E. coli )
Supplier:
VWR International
77750-078
77750-080
77750-082
77750-084
77750-086
77750-078EA
1009.35
CAD
77750-078
77750-080
77750-082
77750-084
77750-086
Human Recombinant EXODUS-2 (from E. coli )
Proteins and Peptides
Exodus-2, also known as CCL21 and 6Ckine, is a chemokine that is strongly produced in the human lymph nodes and spleen. Exodus-2 signals through the chemokine receptor CCR7 to regulate thymocyte and activated T cell migration. Exodus-2 also mediates the homing of lymphocytes to the lymphatic system.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.