Autoclavable Tyvek Breather Bags, Easy Chevron Peel and Self Seal Closure, Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
02430TWBBSSSD 02435TWBBSSSD X0308TWBBSSSD 00510TWBBSSSD 00608TWBBSSSD 00612TWBBSSSD 00628TWBBSSSD 00632TWBBSSSD 00635TWBBSSSD 01424TWBBSSSD 01520TWBBSSSD 01617TWBBSSSD 00206TWBBSSSD 00209TWBBSSSD 00306TWBBSSSD 00308TWBBSSSD 00415TWBBSSSD 00422TWBBSSSD 01011TWBBSSSD 01020TWBBSSSD 01214TWBBSSSD 01217TWBBSSSD 01626TWBBSSSD 01632TWBBSSSD 01635TWBBSSSD 01822TWBBSSSD 02426TWBBSSSD 00713TWBBSSSD 00810TWBBSSSD 00815TWBBSSSD 00818TWBBSSSD 00834TWBBSSSD
77873-624CS 3815.7 CAD
77873-624 77873-484 77873-644 77873-562 77873-616 77873-526 77873-542 77873-640 77873-666 77873-588 77873-606 77873-552 77873-534 77873-570 77873-632 77873-506 77873-584 77873-550 77873-590 77873-600 77873-638 77873-658 77873-636 77873-674 77873-664 77873-620 77873-604 77873-530 77873-536 77873-532 77873-572 77873-576
Autoclavable Tyvek Breather Bags, Easy Chevron Peel and Self Seal Closure, Steam Indicator
Bags Sterilization Pouches
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
  • Manufactured and packaged in an ISO certified cleanroom
  • Chevron peel opening
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Covenient self-sealing closure
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Closure type: Self-seal
Opening: Peel
Delivery information:
Supplied with Certificate of Conformance.