Stopper Tubings, Tyvek/HDPE, Continuous Form
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
024SBT 028SBT 006SBT 012SBT 77873-516 002SBT 003SBT 004SBT 010SBT 77873-582 007SBT 009SBT
77873-592CS 1718.25 CAD
77873-592 77873-628 77873-602 77873-660 77873-516 77873-524 77873-492 77873-520 77873-662 77873-582 77873-538 77873-626
Stopper Tubings, Tyvek/HDPE, Continuous Form
Bags Sterilization Pouches
A significant upgrade over paper-based steam sterilization tubing. Cut tubing to desired length and heat seal both ends closed.
  • Manufactured and packaged in an ISO certified cleanroom
  • Double welded factory seals
  • Recommended for use in conjunction with a secondary Tyvek™ sterilization pouch
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Tubing provides sizing flexibility
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. Tyvek tubing is also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Closure type: Heat seal
Opening: Cut
Delivery information:
Supplied with Certificate of Conformance.